Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Xylene, Reagent Grade, 4 Liter Bottle, 4 per Case
SDP

Supplier:  Research Products International Corp 400460CS

Encompass_Preferred

Colorless liquid
Flammable

Flash Point: 29°C
Boiling Point: 137-140°C

WARNING! This product can expose you to Ethyl benzene, a chemical which is known to the State of California to cause Cancer. For more information go to www.P65Warnings.ca.gov.

Catalog No. 50-998-061


May include imposed supplier surcharges.
Add to Cart

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.