Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Xylene, Reagent Grade, 4 Liters
SDP

Catalog No. 50999353
Encompass_Preferred

Colorless liquid
Flammable

Flash Point: 29°C
Boiling Point: 137-140°C

WARNING! This product can expose you to Ethyl benzene, a chemical which is known to the State of California to cause Cancer. For more information go to www.P65Warnings.ca.gov.

Catalog No. 50-999-353 Supplier Research Products International Corp Supplier No. 400460
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Colorless liquid
Flammable

Flash Point: 29°C
Boiling Point: 137-140°C

WARNING! This product can expose you to Ethyl benzene, a chemical which is known to the State of California to cause Cancer. For more information go to www.P65Warnings.ca.gov.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.