Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylene Cyanol FF, 10g, MP Biomedicals

Catalog No. icn212817 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN212817 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN212817 Supplier MP Biomedicals, Inc Supplier No. 0521281710
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2650-17-1
Molecular Formula C25H27N2NaO6S2
Molecular Weight (g/mol) 538.61
MDL Number MFCD00019481
InChI Key VVLFAAMTGMGYBS-KRQUPCAFSA-M
IUPAC Name sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
SMILES [Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O

Specifications

Quantity 10 g
Chemical Name or Material Xylene Cyanol FF
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.