Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xylene Cyanol FF | 2650-17-1 | 94.69% | 5 G
SDP

Catalog No. 5000409746
Encompass_Preferred

Xylene Cyanol FF is an acid triphenylmethane dye used for histochemical staining of hemoglobin peroxidase or as a tracking dye for DNA sequencing in electrophoresis. It can also be catalyzed by Fe and Al to accelerate oxidation under the addition of double oxidant hydrogen peroxide and potassium periodate, enabling the spectrophotometric determination of Fe and Al in tested solutions. It is also used for denaturing and non-denaturing polyacrylamide gels in DNA tracking.

  • Acid triphenylmethane dye
  • Molecular weight: 538.61
  • Formula: C₂₅H₂₇N₂NaO₆S₂
  • Cas number: 2650-17-1
  • Appearance: Solid
  • Color: Dark green to black
  • Purity: 94.69%
  • Solubility: 10 mg/mL in H₂O (18.57 mM; ultrasonic and warming and heat to 60°C)

Catalog No. 50-004-09746 Supplier Medchemexpress LLC Supplier No. HYD09455G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xylene Cyanol FF is an acid triphenylmethane dye used for histochemical staining of hemoglobin peroxidase or as a tracking dye for DNA sequencing in electrophoresis. It can also be catalyzed by Fe and Al to accelerate oxidation under the addition of double oxidant hydrogen peroxide and potassium periodate, enabling the spectrophotometric determination of Fe and Al in tested solutions. It is also used for denaturing and non-denaturing polyacrylamide gels in DNA tracking.

  • Acid triphenylmethane dye
  • Molecular weight: 538.61
  • Formula: C₂₅H₂₇N₂NaO₆S₂
  • Cas number: 2650-17-1
  • Appearance: Solid
  • Color: Dark green to black
  • Purity: 94.69%
  • Solubility: 10 mg/mL in H₂O (18.57 mM; ultrasonic and warming and heat to 60°C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.