Learn More
Medchemexpress LLC Xylene Cyanol FF | 2650-17-1 | 94.7% | 1 G
SDP
Catalog No. 5000409742
Encompass_Preferred
Xylene Cyanol FF is an acid triphenylmethane dye used for histochemical staining of hemoglobin peroxidase or as a tracking dye for DNA sequencing in electrophoresis. It can also be catalyzed by Fe and Al to accelerate oxidation under the addition of double oxidant hydrogen peroxide and potassium periodate, enabling spectrophotometric determination of Fe and Al in solution.
  • Used for histochemical staining of hemoglobin peroxidase.
  • Tracking dye for DNA sequencing in electrophoresis.
  • Catalyzed by iron and aluminum for spectrophotometric determination in solution.
  • Tracks DNA on different polyacrylamide gels.
  • Suitable for denaturing gel electrophoresis.
Catalog No. 50-004-09742 Supplier Medchemexpress LLC Supplier No. HYD09451G
Xylene Cyanol FF is an acid triphenylmethane dye used for histochemical staining of hemoglobin peroxidase or as a tracking dye for DNA sequencing in electrophoresis. It can also be catalyzed by Fe and Al to accelerate oxidation under the addition of double oxidant hydrogen peroxide and potassium periodate, enabling spectrophotometric determination of Fe and Al in solution.
  • Used for histochemical staining of hemoglobin peroxidase.
  • Tracking dye for DNA sequencing in electrophoresis.
  • Catalyzed by iron and aluminum for spectrophotometric determination in solution.
  • Tracks DNA on different polyacrylamide gels.
  • Suitable for denaturing gel electrophoresis.