Learn More
Abcam x ylene cyanol FF, tracking dye for nucleic acids, 5G
Catalog No. 502704751
Encompass_Preferred
MW 538.61 g/mol. Tracking dye commonly used for nucleic acids. Used in both agarose or polyacrylamide gel electrophoresis. It has a slight negative charge and will migrate in the same direction as DNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4751 Supplier Abcam Supplier No. AB1462965G
MW 538.61 g/mol. Tracking dye commonly used for nucleic acids. Used in both agarose or polyacrylamide gel electrophoresis. It has a slight negative charge and will migrate in the same direction as DNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition.
The product is subject to the following: Abcam Restricted Use Statement