Learn More
Electron Microscopy Sciences EUKITT Neo Mounting Medium 500 ML
SDP
Catalog No. 501928503
Encompass_Preferred
The high-quality classic of mounting media now xylene free. EUKITT Neo is xylene-free and characterized by fast curing, crystal-clear optics, low fluorescence, favorable refractive index, good fluidity and low shrinkage. The preparations are stable without cracking and discoloration. EUKITT Neo is colorless and does not change shape, structure or color of the material to be examined. The unique formulation of this version of EUKITT uses self-sustaining raws that have no solvent base at all. The pleasant odor facilitates working in the laboratory.
Catalog No. 50-192-8503 Supplier Electron Microscopy Sciences Supplier No. 1532220
The high-quality classic of mounting media now xylene free. EUKITT Neo is xylene-free and characterized by fast curing, crystal-clear optics, low fluorescence, favorable refractive index, good fluidity and low shrinkage. The preparations are stable without cracking and discoloration. EUKITT Neo is colorless and does not change shape, structure or color of the material to be examined. The unique formulation of this version of EUKITT uses self-sustaining raws that have no solvent base at all. The pleasant odor facilitates working in the laboratory.