Learn More
Newcomer Supply Xylene, Histology Grade, 1 gallon
SDP
Catalog No. NC2101944
Encompass
Shelf Life is 3 years from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC2101944 Supplier Newcomer Supply Supplier No. 1446C
Shelf Life is 3 years from date of manufacture. Store at 15-30°C.