Learn More
Polysciences, Inc. Xylene, histology grade, 1 gal
SDP
Catalog No. NC1797438
Encompass_Preferred
Routine clearing agent. Also used with Canada balsam in oil-immersion microscopy.
Catalog No. NC1797438 Supplier Polysciences, Inc. Supplier No. 083891
Routine clearing agent. Also used with Canada balsam in oil-immersion microscopy.