Learn More
Ihc World LLC XYLENE, LABORATORY GRADE 1GALL
SDP
Catalog No. NC0692006
Encompass
Xylene, laboratory grade by StatLab. Purified and filtered to deliver a high quality, consistent product. High solvency factor ensures quick and complete clearing. Free of impurities and water. Size: 1 gallon.
Catalog No. NC0692006 Supplier Ihc World LLC Supplier No. 84001
Xylene, laboratory grade by StatLab. Purified and filtered to deliver a high quality, consistent product. High solvency factor ensures quick and complete clearing. Free of impurities and water. Size: 1 gallon.