Learn More
GA International Xylene-Resistant Labels for Paraffin Wax Blocks 1" x 0.25" / 25.4mm x 6.4mm Rectangle White From -55°C to +70°C (-67°F to +158°F)
Supplier:  GA International PRF209C14WH
Encompass_Preferred
Thermal-transfer printable thermoplastic labels that adhere strongly to paraffin wax blocks and other hard-to-stick surfaces such as Teflon, Polytetrafluoroethylene (PTFE), Perfluoroalkoxy (PFA), Parchment paper, etc. ParafiTAG™ labels are water- and chemical-resistant, ideal for identifying paraffin wax blocks for histology applications. A chemical-resistant printout is obtained when printed with our XAR-class of solvent-resistant ribbons. The labels and printed information will resist xylene, toluene, alcohol, and other solvents and chemical exposure without additional protection.
Catalog No. NC1439866