Learn More
Electron Microscopy Sciences Xylene Resistant Thermal Labels, 0.875 x 0.875"
SDP
Catalog No. 5034230
Encompass_Preferred
Pre-cut, peel-off polyester labels for thermal printers labels provide strong solvent resistance, specifically to multiple xylene washes as well as longer term exposure to xylene. Temperature range: -40 degree C to 121 degree C. Xylene labels resist boiling water baths, autoclaving, and moderate freezer temperatures. Rolls are specially top-coated to readily accept thermal transfer printing. Labels are available in a microscope slide size as well as a unique general purpose size for multiple uses.
Note: 100% Virgin polypropylene tubes are recommended. Avoid the use of siliconized surfaces.
Size: 0.875 x 0.875"
Catalog No. 50-342-30 Supplier Electron Microscopy Sciences Supplier No. 7759810
Pre-cut, peel-off polyester labels for thermal printers labels provide strong solvent resistance, specifically to multiple xylene washes as well as longer term exposure to xylene. Temperature range: -40 degree C to 121 degree C. Xylene labels resist boiling water baths, autoclaving, and moderate freezer temperatures. Rolls are specially top-coated to readily accept thermal transfer printing. Labels are available in a microscope slide size as well as a unique general purpose size for multiple uses.
Note: 100% Virgin polypropylene tubes are recommended. Avoid the use of siliconized surfaces.
Size: 0.875 x 0.875"