Learn More
GA International Thermal-transfer, xylene and solvent-resistant resin ribbons 4.33" x 984' / 110mm x 300m Black 1" / 25.4mm
Catalog No. NC1705712
Encompass_Preferred
Xylene and solvent-resistant resin ribbons for thermal-transfer printers. When used with the appropriate label, the ribbon will provide a xylene and chemical-resistant printout. Range of sizes available, ensuring compatibility with most common barcode printer brands.
Catalog No. NC1705712 Supplier GA International Supplier No. XAR110X300C11JZ4
Xylene and solvent-resistant resin ribbons for thermal-transfer printers. When used with the appropriate label, the ribbon will provide a xylene and chemical-resistant printout. Range of sizes available, ensuring compatibility with most common barcode printer brands.