Learn More
Electron Microscopy Sciences EMS Xylene Substitute 1 GL
SDP
Catalog No. 5029800
Encompass_Preferred
The xylene substitute is for use in histology and cytology. It is safer and a more preferable alternative to xylene and cuases minimal tissue shrinkage. It can be used as a solvent and clearing agent, as well as for dissolving paraffin waxes, glues and adhesives. Dries slowly with no residue. It can be used in all procedures that require xylene and is less toxic than xylene. Soluble with alcohol, paraffin embedding media, and mounting media. Biodegradable.
Catalog No. 50-298-00 Supplier Electron Microscopy Sciences Supplier No. 2341001
The xylene substitute is for use in histology and cytology. It is safer and a more preferable alternative to xylene and cuases minimal tissue shrinkage. It can be used as a solvent and clearing agent, as well as for dissolving paraffin waxes, glues and adhesives. Dries slowly with no residue. It can be used in all procedures that require xylene and is less toxic than xylene. Soluble with alcohol, paraffin embedding media, and mounting media. Biodegradable.