Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylene, Vion Biosciences
SCI_ED

Catalog No. S02479 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
4 x 1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02479 500 mL
S02478 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
WARNING: This product can expose you to Ethylbenzene, which is known to the State of California to cause cancer.
Catalog No. S02479 Supplier Aldon Corporation Supplier No. VNXX0020500ML
Add to Cart
Add to Cart
WARNING: This product can expose you to Ethylbenzene, which is known to the State of California to cause cancer.

This xylene is an excellent choice for a clearing and deparaffinizing agent in your histological processes.

  • This xylene is an excellent choice for a clearing and deparaffinizing agent in your histological processes.

Specifications

Boiling Point 139°C to 141°C (283°F to 286°F)
Color Colorless
Content And Storage Store in a cool, dry, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Packaging Type Amber glass bottle
Melting Point Data not available.
Product Line Vion Histology
Product Type Clearing Agent
Quantity 500 mL
Formulation 100% Xylenes
For Use With (Application) Histology
Reagent Type Solvent
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.