Learn More
Electron Microscopy Sciences Xylenes, Reagent A.C.S. 450 ML
SDP
Catalog No. 5029801
Encompass_Preferred
(Xylol, Dimethylbenzene)
C₆H₄(CH₃)₂ F.W. 106.27
This reagent is a mixture of isomers (ortho, meta, and para) and may contain Ethylbenzene.
Catalog No. 50-298-01 Supplier Electron Microscopy Sciences Supplier No. 23400
(Xylol, Dimethylbenzene)
C₆H₄(CH₃)₂ F.W. 106.27
This reagent is a mixture of isomers (ortho, meta, and para) and may contain Ethylbenzene.