Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenol Blue, Indicator Grade, Thermo Scientific™

Catalog No. AC194860010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC194860010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC194860010 Supplier Thermo Scientific Chemicals Supplier No. 194860010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125-31-5
Molecular Formula C23H22O5S
Molecular Weight (g/mol) 410.48
MDL Number MFCD00005870
InChI Key MGUKYHHAGPFJMC-UHFFFAOYSA-N
PubChem CID 67172
IUPAC Name 3,3-bis(4-hydroxy-2,5-dimethylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES CC1=CC(=C(C)C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C=C1C

Specifications

Merck Index 15, 10280
Formula Weight 410.47
Quantity 1 g
Chemical Name or Material Xylenol Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.