Learn More
Specifications
Specifications
| Chemical Name or Material | Xylenol Orange |
| Name Note | 0.2% Alcoholic |
| CAS Max % | 90.8, 4.6, 4.4, 0.2 |
| CAS | 67-63-0, 67-56-1, 3618-43-7 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Specific Gravity | 0.8g/mL |
| Suitability | Passes Test |
| Linear Formula | C31H28N2Na4O13S |
| Decomposition Information | Carbon monoxide; Carbon dioxide |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.