Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenol Orange Disodium Salt, For spectrophotometric determination of metal ions, MilliporeSigma™ Supelco™

Catalog No. 111013283 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3283 5 g
11-101-3282 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3283 Supplier MilliporeSigma Supelco Supplier No. 520975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1611-35-4 | C31H32N2O13S | 672.66 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1611-35-4
Molecular Formula C31H32N2O13S
Molecular Weight (g/mol) 672.66
InChI Key ORZHVTYKPFFVMG-UHFFFAOYSA-N
Synonym 3,3 '-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonephthalein disodium salt
IUPAC Name 2-[({5-[3-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
SMILES CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1

Specifications

Linear Formula C31H30N2Na2O13S
Color Brown to Red
Quantity 5 g
Formula Weight 716.62
Grade Spectrophotometry
Chemical Name or Material Xylenol Orange, Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.