Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenol Orange Indicator, 0.1% (w/v), Ricca Chemical
SDP

Catalog No. R93860001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R93860001A 1 L Natural Poly Bottle
9386-4 120 mL Natural Poly Bottle
9386-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R93860001A Supplier Ricca Chemical Company Supplier No. 938632
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indicator for EDTA Titrations

Chemical Identifiers

CAS 3618-43-7 , 7732-18-5
Molecular Formula C31H28N2Na4O13S

Specifications

Packaging Natural Poly Bottle
Color Orange to Dark Purple
Quantity 1 L
Percent Purity 0.1% W/V
Grade Indicator
Concentration or Composition (by Analyte or Components) 0.1% (w/v) Aqueous Solution
Chemical Name or Material Xylenol Orange
ShelfLife 30 days
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.