Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenol Orange Indicator Mix, Reagents

Catalog No. 01333371
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-371 100 g
01-333-370 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-371 Supplier Reagents Holdings Llc Supplier No. CX300000100C2
Add to Cart
Add to Cart

Xylenol Orange Indicator Mix, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Xylenol Orange Indicator Mix
CAS 7647-14-5, 3618-43-7
Boiling Point 1461°C
Molecular Formula C31H28N2Na4O13S/H2O
Percent Purity 0.1%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 2.1
Color Orange/Red
Quantity 100 g
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.