Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Xylenol Orange, sodium salt, pure, water soluble

Catalog No. AC211220100
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211220100 10 g Glass bottle
AC211220010 1 g Glass bottle
AC211220050 5 g Glass bottle
AC211220500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC211220100 Supplier Thermo Scientific Chemicals Supplier No. 211220100
Add to Cart

CAS: 3618-43-7 | C31H28N2Na4O13S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3618-43-7
Molecular Formula C31H28N2Na4O13S

Specifications

Melting Point 210.0°C
Solubility Information Solubility in water: 510g/L (20°C).
Infrared Spectrum Authentic
Lambda Max 575 to 585nm (0.1N NaOH)
Formula Weight 760.59
Loss on Drying 10.0% (110°C, 1 hr) max.
Grade Pure
Packaging Glass bottle
Suitability (for Zinc titration) Passes test
UV Spectrometry Complies
Color Brown to Brown/Red
Quantity 10 g
Physical Form Crystalline Powder
Chemical Name or Material Xylenol Orange, sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.