Learn More
Specifications
Specifications
| Melting Point | 210°C |
| CAS Max % | 100 |
| Identification | Passes Test |
| Packaging | Amber Glass |
| Suitability | Passes Test |
| Linear Formula | C31H28N2Na4O13S |
| Decomposition Information | Nitrogen oxides; Carbon monoxide; Carbon dioxide |
| Color | Orange to Red |
| Solubility Information | Soluble in water |
| Quantity | 5 g |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Product is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.