Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Xylenol Orange tetrasodium salt, 0.1% w/v aq. soln.

Catalog No. AA42773AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42773AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA42773AE Supplier Thermo Scientific Chemicals Supplier No. 042773AE
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3618-43-7 | C31H28N2Na4O13S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3618-43-7
Molecular Formula C31H28N2Na4O13S
MDL Number MFCD00044293

Specifications

Concentration 0.1% w/v aq. soln.
Color Red
Quantity 100 mL
Physical Form Liquid
Formula Weight 760.59
Chemical Name or Material Xylenol Orange tetrasodium salt
EINECSNumber 222-805-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.