Learn More
Description
Xylenol Orange has been used as a vital stain to study calcification in bone development, injury, and repair. It has been used in the fluorescent histochemical detection of alkaline phosphatase. It has been used in FBX (ferrous benzoic xylenol orange) gel preparation to study the effects of MRI (magnetic resonance imaging) parameters on image quality.
Specifications
Specifications
| Chemical Name or Material | Xylenol Orange Tetrasodium Salt |
| CAS | 3618-43-7 |
| Color | Dark Brown |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 760.58 g/mol |
| Grade | ACS |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.