Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenol Orange Tetrasodium Salt, Indicator for Metal Titration, Solstice

Catalog No. 60046305 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-305 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-046-305 Supplier Solstice Supplier No. 338255G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indicator for metal titration

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3618-43-7
MDL Number MFCD00044293

Specifications

Melting Point 195°C
Packaging Glass Bottle
UN Number NONH for all modes of transport
Color Brown
Quantity 5 g
Physical Form Powder or Crystals
Molecular Weight (g/mol) 760.58
Formula Weight 760.58g/mol
Percent Purity 100%
Chemical Name or Material Xylenol Orange Tetrasodium Salt
RTECSNumber 222-805-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.