Learn More
AdipoGen Xylidyl blue I
SDP
Supplier:  AdipoGen CDXX0009G025
Encompass_Preferred
Chemical. CAS 14936-97-1. Formula C25H20N3NaO6S. MW 513.5. Synthetic Colorimetric reagent for Mg detection. The aqueous solution of XB-1 is red, and turns reddish-violet in the presence of Mg at pH 9 maximum wavelength 510 nm, molar absorptivity 49,000, detection range 0.02-0.4 ppm.
Catalog No. 50-255-9900