Learn More
GA International XyliSTUCK™ – Xylene & Chemical-Resistant Labels for Histology0.866″ x 0.75″ / 22mm x 19.1mm
Catalog No. 502268220
Encompass_Preferred
XyliSTUCK™ labels are specially designed to withstand prolonged xylene immersion, more than 24hrs*, without label detachment or printout deterioration. They are perfectly suited for the most demanding histology (microscope slides) applications that require maximum resistance to various solvents and lab conditions associated with common histology preparation, such as xylene, ethanol gradient series, xylene alternatives, histology stains, and high temperature (70°C) treatments.The permanent adhesive ensures the labels remain affixed, even in the harshest environments. Their advanced design precludes delamination issues and won’t absorb histology stains that can obscure the printout and affect barcode readability. These labels can also withstand exposure to xylene substitutes such as Clearify™, Histo-Clear™, Pro-Par, and others.
This item is not returnable. View return policy
Catalog No. 50-226-8220 Supplier GA International Supplier No. XRM545C11WH
XyliSTUCK™ labels are specially designed to withstand prolonged xylene immersion, more than 24hrs*, without label detachment or printout deterioration. They are perfectly suited for the most demanding histology (microscope slides) applications that require maximum resistance to various solvents and lab conditions associated with common histology preparation, such as xylene, ethanol gradient series, xylene alternatives, histology stains, and high temperature (70°C) treatments.The permanent adhesive ensures the labels remain affixed, even in the harshest environments. Their advanced design precludes delamination issues and won’t absorb histology stains that can obscure the printout and affect barcode readability. These labels can also withstand exposure to xylene substitutes such as Clearify™, Histo-Clear™, Pro-Par, and others.