Learn More
Medchemexpress LLC Xylitol-1-13C | 87-99-0 | 153.14 | 25 MG
SDP
Catalog No. 5000389323
Encompass_Preferred
Xylitol-1-13C is the 13C labeled Xylitol, used as a tracer and an internal standard for quantitative analysis. Stable heavy isotopes of carbon and other elements are incorporated into drug molecules, primarily for quantitation during drug development processes.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of carbon
  • Suitable for drug development process applications
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-89323 Supplier Medchemexpress LLC Supplier No. 25MGHYN0538S
Xylitol-1-13C is the 13C labeled Xylitol, used as a tracer and an internal standard for quantitative analysis. Stable heavy isotopes of carbon and other elements are incorporated into drug molecules, primarily for quantitation during drug development processes.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of carbon
  • Suitable for drug development process applications
  • Potential to affect pharmacokinetic and metabolic profiles of drugs