Learn More
Medchemexpress LLC Xylitol-1-13C | 87-99-0 | MFCD00064292 | 99.0% | 153.14 | C5H12O5 | 10 MG
SDP
Catalog No. 5000296849
Encompass_Preferred
Xylitol-1-13C is the 13C isotopically labeled form of xylitol provided for use as a stable-isotope internal standard and tracer in metabolic, biochemical, and analytical studies. It is supplied at high purity for accurate quantification and tracing in mass spectrometry and NMR workflows.
  • Used as a stable-isotope internal standard for quantitative mass spectrometry
  • Suitable for NMR and metabolic flux analysis
  • High isotopic and chemical purity for accurate tracing
  • Available in small, convenient quantities for analytical workflows
  • Stable at room temperature; follow solvent storage guidelines for extended stability
Catalog No. 50-002-96849 Supplier Medchemexpress LLC Supplier No. HYN0538S10MG
Xylitol-1-13C is the 13C isotopically labeled form of xylitol provided for use as a stable-isotope internal standard and tracer in metabolic, biochemical, and analytical studies. It is supplied at high purity for accurate quantification and tracing in mass spectrometry and NMR workflows.
  • Used as a stable-isotope internal standard for quantitative mass spectrometry
  • Suitable for NMR and metabolic flux analysis
  • High isotopic and chemical purity for accurate tracing
  • Available in small, convenient quantities for analytical workflows
  • Stable at room temperature; follow solvent storage guidelines for extended stability