Learn More
AdipoGen Xylitol
SDP
Catalog No. 502559898
Encompass_Preferred
Chemical. CAS 87-99-0. Formula C5H12O5. MW 152.15. Synthetic. Sugar alcohol used as a sweetener. Unlike other natural or synthetic sweeteners, xylitol is actively beneficial for dental health by reducing caries cavities to a third in regular use and helpful to remineralization. Fair evidence was found that xylitol as chewing gum, lozenges, nasal spray, etc. reduced the incidence of acute middle ear infection in healthy children. Cavity-causing bacteria prefer six-carbon sugars or disaccharides, while xylitol is non-fermentable and cannot be used as an energy source - while still being taken up into the cell due to similar shape and interfering with bacterial growth and reproduction. The harmful micro-organisms are starved in the presence of xylitol, allowing the mouth to remineralize damaged teeth with less interruption. Possessing approximately 33% fewer calories, xylitol is a lower-calorie alternative to table sugar.
Catalog No. 50-255-9898 Supplier AdipoGen Supplier No. CDXX0010G500
Chemical. CAS 87-99-0. Formula C5H12O5. MW 152.15. Synthetic. Sugar alcohol used as a sweetener. Unlike other natural or synthetic sweeteners, xylitol is actively beneficial for dental health by reducing caries cavities to a third in regular use and helpful to remineralization. Fair evidence was found that xylitol as chewing gum, lozenges, nasal spray, etc. reduced the incidence of acute middle ear infection in healthy children. Cavity-causing bacteria prefer six-carbon sugars or disaccharides, while xylitol is non-fermentable and cannot be used as an energy source - while still being taken up into the cell due to similar shape and interfering with bacterial growth and reproduction. The harmful micro-organisms are starved in the presence of xylitol, allowing the mouth to remineralize damaged teeth with less interruption. Possessing approximately 33% fewer calories, xylitol is a lower-calorie alternative to table sugar.