Learn More
Selleck Chemical LLC Xylitol S4546-100mg
SDP
Catalog No. 503110304
Encompass_Preferred
Xylitol (Adonitol Ribitol Xylite D-Xylitol Adonite) is a sugar alcohol derived from XYLOSE by reduction of the carbonyl group It is as sweet as sucrose and used as a noncariogenic sweetener
Catalog No. 50-311-0304 Supplier Selleck Chemical LLC Supplier No. S4546100MG
Xylitol (Adonitol Ribitol Xylite D-Xylitol Adonite) is a sugar alcohol derived from XYLOSE by reduction of the carbonyl group It is as sweet as sucrose and used as a noncariogenic sweetener