Learn More
Medchemexpress LLC Xylitol-5-13C | 87-99-0 | 153.14 | 25 MG
SDP
Catalog No. 5000389562
Encompass_Preferred
Xylitol-5-13C is the 13C labeled Xylit. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89562 Supplier Medchemexpress LLC Supplier No. 25MGHYN0538S2
Xylitol-5-13C is the 13C labeled Xylit. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have potential to affect the pharmacokinetic and metabolic profiles of drugs.