Learn More
Medchemexpress LLC Xylitol-5-13C | 87-99-0 | 153.14 | 5 MG
SDP
Catalog No. 5000388802
Encompass_Preferred
Xylitol-5-13C is the 13C labeled Xylit. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88802 Supplier Medchemexpress LLC Supplier No. HYN0538S25MG
Add to Cart
Add to Cart
Xylitol-5-13C is the 13C labeled Xylit. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.