Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xylitol-5-13C | 87-99-0 | 98.0% | 153.14 g/mol | C4[13C]H12O5 | 10 MG
SDP

Catalog No. 5000296842
Encompass_Preferred

Xylitol-5-13C is a 13C-labeled xylitol isotopic standard supplied as a high-purity reference material for analytical and metabolic studies. It is suitable for use as an internal standard or tracer in mass spectrometry and metabolic flux experiments.

  • Isotopically labeled at the 5-position (13C).
  • Purity 98.0%.
  • Molecular weight 153.14 g/mol.
  • Available in small, pre-weighed quantities, including 10 mg packs.
  • Suitable for quantitative mass spectrometry and tracer studies.

Catalog No. 50-002-96842 Supplier Medchemexpress LLC Supplier No. HYN0538S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xylitol-5-13C is a 13C-labeled xylitol isotopic standard supplied as a high-purity reference material for analytical and metabolic studies. It is suitable for use as an internal standard or tracer in mass spectrometry and metabolic flux experiments.

  • Isotopically labeled at the 5-position (13C).
  • Purity 98.0%.
  • Molecular weight 153.14 g/mol.
  • Available in small, pre-weighed quantities, including 10 mg packs.
  • Suitable for quantitative mass spectrometry and tracer studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.