Learn More
Medchemexpress LLC Xylitol (Standard) | 87-99-0 | MFCD00065074 | 152.15 | 25 MG
SDP
Catalog No. 5000390335
Encompass_Preferred
Xylitol (Standard) is the analytical standard of Xylitol. This product is intended for research and analytical applications. Xylitol can be classified as polyols and sugar alcohols. It is for research use only and not sold to patients.
  • Used as a reference standard for assays
  • Commonly utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-003-90335 Supplier Medchemexpress LLC Supplier No. 25MGHYN0538R
Xylitol (Standard) is the analytical standard of Xylitol. This product is intended for research and analytical applications. Xylitol can be classified as polyols and sugar alcohols. It is for research use only and not sold to patients.
  • Used as a reference standard for assays
  • Commonly utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications