Learn More
Medchemexpress LLC Xylobiose | 6860-47-5 | 1 MG
SDP
Catalog No. 5000421573
Encompass_Preferred
Xylobiose is an orally active Claudin 2/CLDN2 inhibitor and HSP27 inducer that regulates intestinal barrier function and glucose and lipid metabolism. It strengthens intestinal barrier integrity, reduces blood sugar and lipid levels, and mitigates oxidative stress and inflammation. It is used in the study of type 2 diabetes and metabolic syndrome.
  • Orally active Claudin 2/CLDN2 inhibitor and HSP27 inducer
  • Regulates intestinal barrier function
  • Modulates glucose and lipid metabolism
  • Reduces intestinal permeability
  • Enhances cell protection
  • Improves insulin resistance
  • Reduces blood sugar and lipid levels
  • Decreases oxidative stress and inflammatory response
  • Applicable in type 2 diabetes and metabolic syndrome research
Catalog No. 50-004-21573 Supplier Medchemexpress LLC Supplier No. HYN24681MG
Xylobiose is an orally active Claudin 2/CLDN2 inhibitor and HSP27 inducer that regulates intestinal barrier function and glucose and lipid metabolism. It strengthens intestinal barrier integrity, reduces blood sugar and lipid levels, and mitigates oxidative stress and inflammation. It is used in the study of type 2 diabetes and metabolic syndrome.
  • Orally active Claudin 2/CLDN2 inhibitor and HSP27 inducer
  • Regulates intestinal barrier function
  • Modulates glucose and lipid metabolism
  • Reduces intestinal permeability
  • Enhances cell protection
  • Improves insulin resistance
  • Reduces blood sugar and lipid levels
  • Decreases oxidative stress and inflammatory response
  • Applicable in type 2 diabetes and metabolic syndrome research