Learn More
Medchemexpress LLC Xylometazoline-d4 hydrochloride | 1218-35-5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0475S1MG
Encompass_Preferred
Xylometazoline-d4 hydrochloride is a deuterated form of Xylometazoline hydrochloride (HY-B0475). Xylometazoline hydrochloride acts as an α-adrenergic receptor agonist with a Ki ranging from 0.05 to 1.7 μM. It is known for its ability to constrict nasal blood vessels and consequently increase nasal airflow, making it suitable for research concerning nose stuffiness and runny nose.
  • This compound serves as a tracer.
  • It can be utilized as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily for quantitation during the drug development process.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07810