Learn More
Apexbio Technology LLC Xylometazoline HCl 1218-35-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094634
Encompass_Preferred
Xylometazoline hydrochloride (CAS 1218-35-5) is a small molecule -adrenoceptor agonist with pronounced activity at the B-adrenoceptor subtype demonstrating an EC of 99 M Through stimulation of -adrenergic receptors it induces vasoconstriction of nasal mucosal blood vessels which forms the basis for its use as a nasal decongestant Xylometazoline HCl is utilized in biomedical research to investigate -adrenergic receptor signaling pathways and vascular response mechanisms as well as to model sympathomimetic drug action in preclinical studies
Catalog No. 50-000-94634 Supplier Apexbio Technology LLC Supplier No. B20665.1
Xylometazoline hydrochloride (CAS 1218-35-5) is a small molecule -adrenoceptor agonist with pronounced activity at the B-adrenoceptor subtype demonstrating an EC of 99 M Through stimulation of -adrenergic receptors it induces vasoconstriction of nasal mucosal blood vessels which forms the basis for its use as a nasal decongestant Xylometazoline HCl is utilized in biomedical research to investigate -adrenergic receptor signaling pathways and vascular response mechanisms as well as to model sympathomimetic drug action in preclinical studies