Learn More
Medchemexpress LLC Xylometazoline hydrochloride | 1218-35-5 | 99.9% | 1 ML
SDP
Catalog No. 5000407092
Encompass_Preferred
Xylometazoline hydrochloride is an α-adrenergic receptor agonist that constricts nasal blood vessels and increases nasal airflow. It can be used in research for nose stuffiness and runny nose.
  • Alpha-adrenergic receptor agonist
  • Constricts nasal blood vessels
  • Increases nasal airflow
  • Used in research for nose stuffiness and runny nose
  • Ki values for alpha-1A adrenergic receptor: 0.05 μM
  • Ki values for alpha-1B adrenergic receptor: 0.30 μM
  • Ki values for alpha-1D adrenergic receptor: 0.15 μM
  • Ki values for alpha-2A adrenergic receptor: 0.88 μM
  • Ki values for alpha-2B adrenergic receptor: 1.7 μM
  • Ki values for alpha-2C adrenergic receptor: 0.19 μM
Catalog No. 50-004-07092 Supplier Medchemexpress LLC Supplier No. HYB047510MM/1ML
Xylometazoline hydrochloride is an α-adrenergic receptor agonist that constricts nasal blood vessels and increases nasal airflow. It can be used in research for nose stuffiness and runny nose.
  • Alpha-adrenergic receptor agonist
  • Constricts nasal blood vessels
  • Increases nasal airflow
  • Used in research for nose stuffiness and runny nose
  • Ki values for alpha-1A adrenergic receptor: 0.05 μM
  • Ki values for alpha-1B adrenergic receptor: 0.30 μM
  • Ki values for alpha-1D adrenergic receptor: 0.15 μM
  • Ki values for alpha-2A adrenergic receptor: 0.88 μM
  • Ki values for alpha-2B adrenergic receptor: 1.7 μM
  • Ki values for alpha-2C adrenergic receptor: 0.19 μM