Learn More
Medchemexpress LLC Xylometazoline (hydrochloride) | 1218-35-5 | 99.9% | 10 G
SDP
Catalog No. 5000407940
Encompass_Preferred
Xylometazoline hydrochloride is an alpha-adrenergic receptor agonist that constricts nasal blood vessels and increases nasal airflow. It is used in nose stuffiness and runny nose research.
  • Alpha-adrenergic receptor agonist (Ki=0.05-1.7 μM)
  • Constricts nasal blood vessels
  • Increases nasal airflow
  • Suitable for nose stuffiness research
  • Suitable for runny nose research
  • Shows affinity for α1A-adrenoceptor and α2B-adrenoceptor in HEK293 cells
  • Improves sinusitis in rats in vivo
Catalog No. 50-004-07940 Supplier Medchemexpress LLC Supplier No. HYB047510G
Xylometazoline hydrochloride is an alpha-adrenergic receptor agonist that constricts nasal blood vessels and increases nasal airflow. It is used in nose stuffiness and runny nose research.
  • Alpha-adrenergic receptor agonist (Ki=0.05-1.7 μM)
  • Constricts nasal blood vessels
  • Increases nasal airflow
  • Suitable for nose stuffiness research
  • Suitable for runny nose research
  • Shows affinity for α1A-adrenoceptor and α2B-adrenoceptor in HEK293 cells
  • Improves sinusitis in rats in vivo