Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Xylometazoline hydrochloride, 97+%

Catalog No. AC461990050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461990050 5 g
AC461990250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461990050 Supplier Thermo Scientific Chemicals Supplier No. 461990050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1218-35-5 | C16H24N2·HCL | 280.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218-35-5
Molecular Formula C16H24N2·HCL
Molecular Weight (g/mol) 280.84
InChI Key YGWFCQYETHJKNX-UHFFFAOYSA-N
Synonym xylometazoline hydrochloride, xylometazoline hcl, 2-4-tert-butyl-2,6-dimethylbenzyl-2-imidazoline hydrochloride, unii-x5s84033nz, dsstox_cid_25223, dsstox_rid_80760, dsstox_gsid_45223, 2-4-tert-butyl-2,6-dimethyl-phenyl methyl-4,5-dihydro-1h-imidazole hydrochloride, 2-4-tert-butyl-2,6-dimethylphenyl methyl-4,5-dihydro-1h-imidazole hydrochloride
PubChem CID 5282386
IUPAC Name 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole;hydrochloride
SMILES CC1=CC(=CC(=C1CC2=NCCN2)C)C(C)(C)C.Cl

Specifications

Quantity 5 g
Solubility Information (5% in water) Clear colorless to light yellow
Formula Weight 280.84
Percent Purity 98%
Chemical Name or Material Xylometazoline hydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 214-936-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.