Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylometazoline Hydrochloride, Spectrum™ Chemical
SDP

Catalog No. 18607784 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-784 1 g Amber Glass Bottle
18-607-785 5 g Amber Glass Bottle
18-607-786 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-784 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. X20011GM
Add to Cart
Add to Cart

X2001, 1218-35-5, C16H24N2·HCl

Spectrum™ Chemical Xylometazoline Hydrochloride , also known as xylomethazoline, is used topically as a nasal decongestant in either drops or sprays. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 1218-35-5
Molecular Formula C16H25ClN2
Molecular Weight (g/mol) 280.84
InChI Key YGWFCQYETHJKNX-UHFFFAOYSA-N
IUPAC Name hydrogen 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole chloride
SMILES [H+].[Cl-].CC1=CC(=CC(C)=C1CC1=NCCN1)C(C)(C)C

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 g
UN Number UN2811
Percent Purity 98%
Grade Reagent
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.