Learn More
Medchemexpress LLC Xylose | 58-86-6 | MFCD00064360 | 150.13 | 500 G
SDP
Supplier:  Medchemexpress LLC HYN0537500G
Encompass_Preferred
Xylose (D-(+)-Xylose) is a natural pentose sugar. It is catalyzed by xylose isomerase to form xylulose, which is a key step in the anaerobic ethanol fermentation of Xylose. It provides substances and energy for cells, acting as a carbon source for the biosynthesis of high-value chemicals and biofuel.
  • Can be used by microorganisms to produce fuels, chemicals, and bulk industrial enzymes.
  • Suitable for exploring lignocellulose resources.
  • Offers a new direction for microbial fermentation.
Catalog No. 50-003-88568