Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylosucrose 98.0+%, TCI America™
SDP

Catalog No. X00651G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00651G 1 g
X0065100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00651G Supplier TCI America Supplier No. X00651G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 512-66-3 | C11H20O10 | 312.271 g/mol

Chemical Identifiers

CAS 512-66-3
Molecular Formula C11H20O10
Molecular Weight (g/mol) 312.271
InChI Key DRYHSZUFKNRFCT-ROIIQKICSA-N
Synonym beta-D-Fructofuranosyl alpha-D-Xylopyranoside
PubChem CID 53384542
IUPAC Name (2R,3S,4S,5R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxyoxane-3,4,5-triol
SMILES C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O

Specifications

Melting Point 114°C
Color White
Quantity 1 g
Formula Weight 312.27
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Xylosucrose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.