Learn More
Aviva Systems Biology Corp XYLT1 Antibody - middle region (ARP49700_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49700_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XYLT1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human XYLT1. Alternate names: PXYLT1, XT-I, XT1, XTI, XYLTIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-444