Learn More
Aviva Systems Biology Corp XYLT2 Antibody - middle region (ARP49702_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49702_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XYLT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human XYLT2. Alternate names: XT-II, XT2, xylT-II, PXYLT2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60148