Learn More
Iwai North America Xyltech™ MSC-01: Human Mesenchymal Stem Cell (MSC) Proliferation Control Medium. Size: 100mL
Catalog No. NC2733038
Encompass_Preferred
Xyltech™ MSC-01 is serum-free novel culture medium that suppresses cell growth while maintaining cell morphology and the ability to differentiate optimized for human mesenchymal stem cells (MSC). This product is perfect for those looking to slow cell growth to prevent over-confluency
Catalog No. NC2733038 Supplier Iwai North America Supplier No. B10401
Xyltech™ MSC-01 is serum-free novel culture medium that suppresses cell growth while maintaining cell morphology and the ability to differentiate optimized for human mesenchymal stem cells (MSC). This product is perfect for those looking to slow cell growth to prevent over-confluency