Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XYLYX BIO INC Xylyx Bio Hydrogel Kit, Kidney dECM, 1 mL, Porcine

Catalog No. NC3992202
Encompass

The TissueSpec™ Kidney dECM Hydrogel Kit provides a physiologically relevant kidney matrix for advanced cell culture. This lot-to-lot consistent kit offers 1 mL at 6 mg/mL for various 3D cell culture studies.

  • Kit includes 2 x 0.3 mL TissueSpec™ Kidney dECM, 1 x 1 mL Component A, and 1 x 1 mL Component B.
  • Ensures >95% viability with human primary fibroblasts and is certified sterile.
  • Optimal working concentration of 6 mg/mL with a neutral pH.
  • Suitable for disease modeling, drug testing, organ-on-chip, organoids, spheroids, and stem cell research.
  • Derived from porcine kidney with a 12-month shelf life at 2–8°C.
  • Validated with various cell types, multi-well formats, and imaging modalities.
  • Gelation time is 30 minutes at 37°C.

Catalog No. NC3992202 Supplier XYLYX BIO INC Supplier No. MTSKY101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The TissueSpec™ Kidney dECM Hydrogel Kit provides a physiologically relevant kidney matrix for advanced cell culture. This lot-to-lot consistent kit offers 1 mL at 6 mg/mL for various 3D cell culture studies.

  • Kit includes 2 x 0.3 mL TissueSpec™ Kidney dECM, 1 x 1 mL Component A, and 1 x 1 mL Component B.
  • Ensures >95% viability with human primary fibroblasts and is certified sterile.
  • Optimal working concentration of 6 mg/mL with a neutral pH.
  • Suitable for disease modeling, drug testing, organ-on-chip, organoids, spheroids, and stem cell research.
  • Derived from porcine kidney with a 12-month shelf life at 2–8°C.
  • Validated with various cell types, multi-well formats, and imaging modalities.
  • Gelation time is 30 minutes at 37°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.