Learn More
Enterprise Technology Solutions Xyron Non-Heat Laminator, Portable, Black/Chrome
SDP
Catalog No. 5000740392
Encompass_Preferred
Portable cold laminator does not require any electricity, batteries or warm-up time to ensure it's ready when you need it. The cold laminate is photo-safe, allowing you to work without the worries of heat or toxic fumes. Easy drop-in cartridges allow you to make quick changes between projects. High-quality laminate will not peel for long-lasting protection. To laminate, simply slip your documents into the entry tray, crank the handle and trim with the attached trimmer. Flexible design is compatible with multiple refill options (sold separately) to accommodate a variety of projects.
Catalog No. 50-007-40392 Supplier Enterprise Technology Solutions Supplier No. XRN624672
Portable cold laminator does not require any electricity, batteries or warm-up time to ensure it's ready when you need it. The cold laminate is photo-safe, allowing you to work without the worries of heat or toxic fumes. Easy drop-in cartridges allow you to make quick changes between projects. High-quality laminate will not peel for long-lasting protection. To laminate, simply slip your documents into the entry tray, crank the handle and trim with the attached trimmer. Flexible design is compatible with multiple refill options (sold separately) to accommodate a variety of projects.